siRNA Details
Gene Symbol: 4632404H12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040741.1 | - | 1 | 1566 | ||
AK040882.1 | - | 1 | 1594 | |||
AK047674.1 | - | 1 | 1583 | |||
AK048101.1 | - | 1 | 1752 | |||
AK084338.1 | - | 1 | 1584 | |||
AK164170.1 | - | 1 | 1643 | |||
AK169469.1 | - | 1 | 1591 | |||
BC063049.1 | - | 1 | 1143 | |||
BC080683.1 | - | 1 | 1582 |
