siRNA Details
Gene Symbol: 2700063P09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012482.1 | - | 1 | 86 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY711609.1 | - | 1 | 86 | Chr. 4 - 57611535 - 57611872 [+] on Build GRCm38 |
