siRNA Details
Gene Symbol: 4930430J02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040729.1 | 1 | 109 | |||
GenBank mRNA | AK015253.1 | - | 1 | 110 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714811.1 | - | 1 | 110 | Chr. 9 - 57391070 - 57400371 [+] on Build GRCm38 |
