siRNA Details
Gene Symbol: 4930505G20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045761.1 | 1, 2 | 39 | |||
GenBank mRNA | AK015702.1 | - | 1, 2 | 41 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715244.1 | - | 1, 2 | 40 | Chr. 14 - 115396550 - 115409552 [-] on Build GRCm38 |
