siRNA Details
Gene Symbol: 4930505G20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045761.1 | 4 | 549 | |||
GenBank mRNA | AK015702.1 | - | 4 | 551 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930505G20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045761.1 | 4 | 549 | |||
GenBank mRNA | AK015702.1 | - | 4 | 551 |