siRNA Details
Gene Symbol: 2310047K21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009882.1 | - | 1 | 1462 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CD770368.1 | - | 1 | 508 | Chr. 2 - 104817875 - 104818493 [+] on Build GRCm38 |
