siRNA Details
Gene Symbol: 1700120G07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046050.1 | 1, 2 | 33 | |||
GenBank mRNA | AK007216.1 | - | 1, 2 | 34 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707307.1 | - | 1, 2 | 37 | Chr. 7 - 135191765 - 135196002 [-] on Build GRCm38 |
