siRNA Details
Gene Symbol: D630010B17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045629.1 | 3 | 224 | |||
GenBank mRNA | AK085312.1 | - | 3 | 225 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB660866.1 | - | 3 | 222 | Chr. 15 - 94247204 - 94254388 [+] on Build GRCm38 |
