siRNA Details
Gene Symbol: D630010B17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045629.1 | 4 | 982 | |||
GenBank mRNA | AK085312.1 | - | 4 | 983 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB497702.2 | - | 2 | 569 | Chr. 15 - 94254197 - 94255606 [+] on Build GRCm38 |
