siRNA Details
Gene Symbol: Gm31452 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK172672.1 | - | 3, 4 | 512 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY766092.1 | - | 1, 2 | 361 | Chr. 13 - 107085990 - 107087797 [+] on Build GRCm38 |
