siRNA Details
Gene Symbol: Lnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038203.1 | - | 718 | |||
AK147232.1 | - | 6123 | ||||
AK153220.1 | - | 2458 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038203.1 | - | 718 | |||
AK147232.1 | - | 6123 | ||||
AK153220.1 | - | 2458 |