siRNA Details
Gene Symbol: Lnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038203.1 | - | 139 | |||
AK147232.1 | - | 5544 | ||||
AK153220.1 | - | 1879 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CF723060.1 | - | 1 | 123 | Chr. 17 - 17522499 - 17523206 [-] on Build GRCm38 |
