siRNA Details
Gene Symbol: Ikzf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020006.1 | - | 1 | 499 | ||
AK036657.1 | - | 3 | 1778 | |||
AK080208.1 | - | 1 | 1004 | |||
AK157177.1 | - | 3 | 1689 | |||
AK169802.1 | - | 1 | 2120 |
