siRNA Details
Gene Symbol: Gm33104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029599.1 | - | 3 | 368 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB614960.1 | - | 3 | 368 | Chr. 10 - 19486715 - 19507243 [+] on Build GRCm38 |
