siRNA Details
Gene Symbol: March3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018160.1 | - | 1 | 1464 | ||
AK029887.1 | - | 1 | 1453 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DT908940.1 | - | 1 | 125 | Chr. 18 - 56923475 - 56924220 [-] on Build GRCm38 |
