siRNA Details
Gene Symbol: 5730480H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045500.1 | 4 | 1688 | |||
GenBank mRNA | AK017706.1 | - | 4 | 1689 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV024914.2 | - | 4 | 969 | Chr. 5 - 48396345 - 48414265 [+] on Build GRCm38 |
