siRNA Details
Gene Symbol: 5730478J17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017704.1 | - | 1 | 1234 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CN670191.1 | - | 1 | 102 | Chr. 18 - 72156997 - 72157499 [-] on Build GRCm38 |
