siRNA Details
Gene Symbol: Gm26535 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042044.1 | - | 3 | 197 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB638824.1 | - | 3 | 197 | Chr. 10 - 37337486 - 37377686 [+] on Build GRCm38 |
