siRNA Details
Gene Symbol: 1700016K05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006039.1 | - | 2 | 277 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706157.1 | - | 2 | 272 | Chr. 17 - 66714459 - 66720485 [+] on Build GRCm38 |
