siRNA Details
Gene Symbol: Gm20556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040347.1 | 3 | 776 | |||
GenBank mRNA | AK079813.1 | - | 3 | 777 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm20556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040347.1 | 3 | 776 | |||
GenBank mRNA | AK079813.1 | - | 3 | 777 |