siRNA Details
Gene Symbol: 9130227L01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045837.1 | 3 | 320 | |||
GenBank mRNA | AK079812.1 | - | 3 | 320 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB635543.1 | - | 3 | 320 | Chr. 1 - 55931241 - 55957925 [+] on Build GRCm38 |
