siRNA Details
Gene Symbol: Rcor3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035958.1 | - | 1 | 2712 | ||
AK053482.1 | - | 1 | 945 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB539863.2 | - | 1 | 247 | Chr. 1 - 192128108 - 192128750 [-] on Build GRCm38 |
