siRNA Details
Gene Symbol: Gm12916 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079012.1 | - | 4 | 772 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV381235.2 | - | 2 | 520 | Chr. 4 - 124293691 - 124296415 [-] on Build GRCm38 |
