siRNA Details
Gene Symbol: Gm13571 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK050909.1 | - | 456 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB654683.1 | - | 2 | 456 | Chr. 2 - 60338188 - 60355769 [+] on Build GRCm38 |
