siRNA Details
Gene Symbol: Gm34471 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041352.1 | - | 2 | 208 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB638018.1 | - | 2 | 208 | Chr. 13 - 113532029 - 113567767 [-] on Build GRCm38 |
