siRNA Details
Gene Symbol: I730030J21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045781.1 | 1 | 158 | |||
NR_045782.1 | 1 | 158 | ||||
GenBank mRNA | AK146183.1 | - | 1 | 159 | ||
AK163102.1 | - | 1 | 142 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: I730030J21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045781.1 | 1 | 158 | |||
NR_045782.1 | 1 | 158 | ||||
GenBank mRNA | AK146183.1 | - | 1 | 159 | ||
AK163102.1 | - | 1 | 142 |