siRNA Details
Gene Symbol: BC048602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045280.1 | 2 | 358 | |||
GenBank mRNA | BC048602.1 | - | 2 | 358 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU946253.1 | - | 2 | 361 | Chr. 15 - 35307020 - 35328537 [-] on Build GRCm38 |
