siRNA Details
Gene Symbol: Gm3289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040992.1 | - | 605 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB219806.2 | - | 2 | 538 | Chr. 6 - 21852983 - 21858112 [-] on Build GRCm38 |
