siRNA Details
Gene Symbol: Gm31456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133121.1 | - | 609 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB019023.2 | - | 1 | 124 | Chr. 14 - 38318387 - 38337126 [+] on Build GRCm38 |
