siRNA Details
Gene Symbol: 4921520J07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014930.1 | - | 1 | 37 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714446.1 | - | 1 | 35 | Chr. 7 - 128522563 - 128523148 [-] on Build GRCm38 |
