siRNA Details
Gene Symbol: 2610029K11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011616.1 | - | 1 | 555 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ306777.1 | - | 1 | 40 | Chr. 17 - 34628340 - 34628760 [+] on Build GRCm38 |
