siRNA Details
Gene Symbol: 1700127F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018992.1 | - | 1 | 157 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707401.1 | - | 1 | 157 | Chr. 12 - 112875211 - 112875814 [-] on Build GRCm38 |
