siRNA Details
Gene Symbol: 1700129O19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018999.1 | - | 1 | 253 | ||
BM942913.1 | - | 1 | 128 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU945787.1 | - | 1 | 268 | Chr. 13 - 49340848 - 49341392 [-] on Build GRCm38 |
