siRNA Details
Gene Symbol: 4932407I05 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029928.1 | - | 2 | 3455 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV273145.2 | - | 1 | 119 | Chr. 17 - 28855147 - 28855744 [-] on Build GRCm38 |
