siRNA Details
Gene Symbol: 4930595O18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016395.1 | - | 1 | 597 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716047.1 | - | 1 | 597 | Chr. 6 - 100922550 - 100923156 [-] on Build GRCm38 |
