siRNA Details
Gene Symbol: 4930594O21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016393.1 | - | 2 | 84 | ||
BB617223.1 | - | 2 | 93 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CN843869.1 | - | 2 | 68 | Chr. 3 - 37609889 - 37614813 [+] on Build GRCm38 |
