siRNA Details
Gene Symbol: 2900035I09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013616.1 | - | 1 | 613 | ||
AK032244.1 | - | 1 | 1670 | |||
AV150944.1 | - | 1 | 97 | |||
AV152299.1 | - | 1 | 228 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF471619.1 | - | 1 | 450 | Chr. 7 - 29890849 - 29891368 [-] on Build GRCm38 |
