siRNA Details
Gene Symbol: 2900035I09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013616.1 | - | 1 | 142 | ||
AK032244.1 | - | 1 | 1199 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712580.1 | - | 1 | 139 | Chr. 7 - 29890911 - 29891527 [-] on Build GRCm38 |
