siRNA Details
Gene Symbol: 4930471C04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046192.1 | 3 | 239 | |||
GenBank mRNA | AK019642.1 | - | 3 | 239 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715092.1 | - | 3 | 239 | Chr. 14 - 64099601 - 64104235 [-] on Build GRCm38 |
