siRNA Details
Gene Symbol: 4930408O17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015114.1 | - | 2 | 205 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714660.1 | - | 2 | 205 | Chr. 12 - 104865330 - 104871525 [+] on Build GRCm38 |
