siRNA Details
Gene Symbol: 4930412B13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040631.1 | 1 | 738 | |||
GenBank mRNA | AK015116.1 | - | 1 | 738 | ||
AK029598.1 | - | 1 | 738 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV260150.2 | - | 1 | 454 | Chr. 2 - 117822212 - 117909134 [+] on Build GRCm38 |
