siRNA Details
Gene Symbol: Fam155a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK149226.1 | - | 4 | 458 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY742096.1 | - | 4 | 458 | Chr. 8 - 9430007 - 9770104 [-] on Build GRCm38 |
