siRNA Details
Gene Symbol: E130102C15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021367.1 | - | 1 | 1107 | ||
AK081928.1 | - | 1 | 1281 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB377026.2 | - | 1 | 595 | Chr. X - 101131629 - 101132255 [-] on Build GRCm38 |
