siRNA Details
Gene Symbol: 6430519N07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032319.1 | - | 1 | 2056 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV341187.2 | - | 1 | 132 | Chr. 6 - 21235523 - 21236156 [+] on Build GRCm38 |
