siRNA Details
Gene Symbol: LOC102641455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK146407.1 | - | 2 | 507 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY738567.1 | - | 2 | 504 | Chr. 17 - 28522532 - 28523146 [-] on Build GRCm38 |
