siRNA Details
Gene Symbol: Gm31452 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK143700.1 | - | 2 | 624 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY732453.1 | - | 2 | 621 | Chr. 13 - 107063695 - 107098412 [+] on Build GRCm38 |
