siRNA Details
Gene Symbol: 3010001F23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045451.1 | 4 | 585 | |||
NR_045452.1 | 5 | 566 | ||||
GenBank mRNA | AK013876.1 | - | 4 | 588 | ||
EL960655.1 | - | 2 | 157 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712834.1 | - | 4 | 584 | Chr. X - 152368573 - 152416526 [+] on Build GRCm38 |
