siRNA Details
Gene Symbol: 1700052M09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006774.1 | - | 1 | 525 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706757.1 | - | 1 | 528 | Chr. 14 - 29840855 - 29841725 [+] on Build GRCm38 |
