siRNA Details
Gene Symbol: Gm35234 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK044087.1 | - | 384 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB274645.2 | - | 2 | 234 | Chr. 18 - 59825432 - 59855440 [-] on Build GRCm38 |
