siRNA Details
Gene Symbol: 7330423F06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020185.1 | - | 1 | 221 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY719600.1 | - | 1 | 221 | Chr. 5 - 8831147 - 8831465 [+] on Build GRCm38 |
